
Methyl-4-(cyanomethyl)benzoate 97+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76469-88-0
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00060695
:175.19 g/mol
Boiling Pt:110 °C/0.23mmHg
Melting Pt:55…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together