exo-Hexahydro-1H-4,7-methanoisoindole-1,3(2H)-dione 95%
Catalog # APOSOR46129-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
:165.1900 g/mol
Storage Temperature:Ambient
CAS Number:14805-29-9
Formula:C₉H₁₁NO₂
MDL Number:MFCD12964181