Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Dimethylamino)nicotinaldehyde
6-(Dimethylamino)nicotinaldehyde
6-(Dimethylamino)nicotinaldehyde
APOSOR46111-1G
:
Apollo Scientific
:
6-(Dimethylamino)nicotinaldehyde
APOSOR46111-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
150.18 g/mol
:
51…53 °C
:
GHS07
:
Ambient
:
149805-92-5
:
C₈H₁₀N₂O
:
MFCD08272100