
5-Methoxy-2-methylindole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1076-74-0
:
Formula:C₁₀H₁₁NO
MDL Number:MFCD00005620
Melting Pt:85…88 °C
:161.2 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together