Order Entry
Czech Republic
ContactUsLinkComponent
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Irritant
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Catalog # APOSOR46059-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-5H-pyrrolo[3,2-d]pyrimidine
Catalog # APOSOR46059-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.5691 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:84905-80-6
Formula:C₆H₄ClN₃
MDL Number:MFCD06658411