
(R)-3-Aminotetrahydrofuran hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1072015-52-1
:
Formula:C₄H₁₀ClNO
MDL Number:MFCD08461720
:123.5800 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together