
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1312765-17-5
:
Formula:C₁₀H₁₆BNO₃S
MDL Number:MFCD10688611
:241.12 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together