
4-tert-Butylpiperidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69682-13-9
:
Formula:C₉H₂₀ClN
MDL Number:MFCD05865119
:177.72 g/mol
Melting Pt:302…303 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together