Order Entry
Czech Republic
ContactUsLinkComponent
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
Catalog # APOSOR4578-25G
Supplier:  Apollo Scientific
CAS Number:  
(R)-(+)-3-Hydroxypiperidine hydrochloride 98%
Catalog # APOSOR4578-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :137.607 g/mol
Melting Pt:195…196 °C
Storage Temperature:Ambient
CAS Number:198976-43-1
Formula:C₅H₁₂ClNO
MDL Number:MFCD00192221