

2-(p-Toluenesulphonyl)acetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:31378-03-7
:
Formula:C₁₅H₁₄O₃S
MDL Number:MFCD00026002
:274.340 g/mol
Melting Pt:109…110 °C
EINECS:250-605-0
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together