
4-Bromo-1,3-dimethoxybenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17715-69-4
:
Formula:C₈H₉BrO₂
MDL Number:MFCD00009844
:217.06 g/mol
Boiling Pt:153…155 °C/18mmHg
Melting Pt:24…26 °C
Density:1.507 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:241-717-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together