Order Entry
Czech Republic
ContactUsLinkComponent
 
1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride
Catalog # APOSOR451136-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Pyrrolidin-3-yl-1H-pyrazole dihydrochloride
Catalog # APOSOR451136-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.11 g/mol
Storage Temperature:Ambient
CAS Number:1242339-08-7
Formula:C₇H₁₃Cl₂N₃
MDL Number:MFCD16039347