2-(4-Methylpiperazin-1-ylmethyl)benzoic acid
Catalog # APOSOR451094-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:234.2900 g/mol
Storage Temperature:Ambient
CAS Number:514209-40-6
Formula:C₁₃H₁₈N₂O₂
MDL Number:MFCD06797777