
2,6-Dichloro-3-cyanopyrazine
Catalog # APOSOR45109-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:173.9875 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:313339-92-3
Formula:C₅HCl₂N₃
MDL Number:MFCD13193460