(R)-3-Phenylmorpholine hydrochloride
Catalog # APOSOR451060-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.68 g/mol
Storage Temperature:Ambient
CAS Number:1624261-27-3
Formula:C₁₀H₁₃NO·ClH
MDL Number:MFCD12910599