Order Entry
Czech Republic
ContactUsLinkComponent
4-(Pyrrolidin-2-ylmethyl)morpholine
4-(Pyrrolidin-2-ylmethyl)morpholine
Catalog # APOSOR451052-5G
Supplier:  Apollo Scientific
CAS Number:  
4-(Pyrrolidin-2-ylmethyl)morpholine
Catalog # APOSOR451052-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :170.26 g/mol
Storage Temperature:Ambient
CAS Number:215503-90-5
Formula:C₉H₁₈N₂O
MDL Number:MFCD06653314