1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride
Catalog # APOSOR451051-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:243.18 g/mol
Storage Temperature:Ambient
CAS Number:790223-65-3
Formula:C₉H₂₀Cl₂N₂O
MDL Number:MFCD11858471