3-(4-Bromophenoxy)azetidine hydrochloride
Catalog # APOSOR451022-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:264.56 g/mol
Storage Temperature:Ambient
CAS Number:1380571-76-5
Formula:C₉H₁₁BrClNO
MDL Number:MFCD05664840