
tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:190900-21-1
:
Formula:C₁₀H₁₈N₂O₃
MDL Number:MFCD04115319
:214.2615 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together