Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
2-Phenoxyethanol

2-Phenoxyethanol

Irritant
Supplier:   Apollo Scientific
2-Phenoxyethanol
Supplier:  Apollo Scientific
Certificates
2-Phenoxyethanol

3 Options Available Below

Product Details & Documents
Synonyms:  ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
  • CAS Number:122-99-6
  • Formula:C₆H₅OCH₂CH₂OH
  • MDL Number:MFCD00002857
  • Molecular Weight:138.17 g/mol
  • Boiling Pt:237…246 °C
  • Melting Pt:41944 °C
  • Density:1.107 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:204-589-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 615,00 Kč
    615,00 Kč
    1 * 25 g
       615,00 Kč

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 697,00 Kč
    697,00 Kč
    1 * 500 g
       697,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 400,00 Kč
    1 400,00 Kč
    1 * 2,5 kg
       1 400,00 Kč

    Specifications:
    New Product
    New Product