Order Entry
Czech Republic
ContactUsLinkComponent
1-Methylimidazole-5-carboxylic acid methyl ester 97%
Irritant
1-Methylimidazole-5-carboxylic acid methyl ester 97%
Catalog # APOSOR4390-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Methylimidazole-5-carboxylic acid methyl ester 97%
Catalog # APOSOR4390-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:115…117 °C/8mmHg
Melting Pt:56…57 °C
 :140.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:17289-20-2
Formula:C₆H₈N₂O₂
MDL Number:MFCD01567300