
5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:182415-24-3
:
Formula:C8H6N2O2S
MDL Number:MFCD03030176
:194.2104 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together