
1-Bromo-2-naphthol 98%
Catalog # APOSOR43657-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.07 g/mol
Melting Pt:78…81 °C
:GHS07
Storage Temperature:Ambient
CAS Number:573-97-7
Formula:C₁₀H₇BrO
MDL Number:MFCD00003869