
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76189-55-4
:
Formula:C₄₄H₃₂P₂
MDL Number:MFCD00010805
Melting Pt:239…242 °C
:622.69 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together