1-Chloro-2-methyl-2-propanol 97%
Catalog # APOSOR4348-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:108.57 g/mol
Boiling Pt:124…125 °C
Density:1.05 g/cm³ (25 °C)
Flash Pt:37 °C
Storage Temperature:Ambient
CAS Number:558-42-9
Formula:C₄H₉ClO
MDL Number:MFCD00021807