Order Entry
Czech Republic
ContactUsLinkComponent
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole 95%
Irritant
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole 95%
Catalog # APOSOR42179-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6,7-Dihydro-5H-pyrrolo[1,2-a]imidazole 95%
Catalog # APOSOR42179-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :108.14 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:59646-16-1
Formula:C₆H₈N₂
MDL Number:MFCD11870771