
(R)-Methyl-2-((tert-butoxycarbonyl)amino)-3-iodopropanoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:93267-04-0
:
Formula:C₉H₁₆INO₄
MDL Number:MFCD00216579
Melting Pt:50…52 °C
:329.13 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together