Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-chloro-5-methylpyridine
2-Bromo-3-chloro-5-methylpyridine
2-Bromo-3-chloro-5-methylpyridine
APOSOR42133-1G
:
Apollo Scientific
:
2-Bromo-3-chloro-5-methylpyridine
APOSOR42133-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
206.48 g/mol
:
Ambient
:
65550-81-4
:
C₆H₅BrClN
:
MFCD09839259