Order Entry
Czech Republic
ContactUsLinkComponent
Decahydronaphthalene (mixture of cis and trans isomers) 98%
Flammable
Corrosive
Toxic
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) 98%
Catalog # APOSOR42013-2.5L
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Toxic
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) 98%
Catalog # APOSOR42013-2.5L
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :138.25 g/mol
Boiling Pt:189…191 °C
Melting Pt:–125 °C
Density:0.896 g/cm³ (25 °C)
Flash Pt:57.2 °C
Storage Temperature:Ambient
EINECS:202-046-9
UN:1147
ADR:3,III
 :GHS02
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:91-17-8
Formula:C₁₀H₁₈
MDL Number:MFCD00004130