Order Entry
Czech Republic
ContactUsLinkComponent
N-Ethyl-4-boronobenzenesulphonamide 97%
Irritant
N-Ethyl-4-boronobenzenesulphonamide 97%
Catalog # APOSOR4132-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-Ethyl-4-boronobenzenesulphonamide 97%
Catalog # APOSOR4132-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.06 g/mol
Melting Pt:132…136 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:871329-65-6
Formula:C₈H₁₂BNO₄S
MDL Number:MFCD07363751