
N-Isopropyl 4-boronobenzenesulphonamide 98%
Catalog # APOSOR4127-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:243.08 g/mol
Melting Pt:136…140 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:850589-31-0
Formula:C₉H₁₄BNO₄S
MDL Number:MFCD07363752