
4-(2-Methoxyethylcarbamoyl)phenylboronic acid 98%
Catalog # APOSOR4125-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:223.04 g/mol
Melting Pt:120…124 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:850589-34-3
Formula:C₁₀H₁₄BNO₄
MDL Number:MFCD04115682