
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97%
Catalog # APOSOR41001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.13 g/mol
Melting Pt:57…62 °C
:GHS07
Storage Temperature:Ambient
CAS Number:885692-91-1
Formula:C₁₁H₁₇BO₂S
MDL Number:MFCD08063137