2-Aminophenol 99%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol
CAS Number:95-55-6
:
Formula:C₆H₇NO
MDL Number:MFCD00007690
Boiling Pt:235
Molecular Weight:109.13 g/mol
Melting Pt:170…175 °C
Flash Pt:168 °C
ADR:6.1 III
Merck Index:13,00460
EINECS:202-431-1
UN:2512
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together