



tert-Butylchlorodimethylsilane 50% in toluene
Catalog # APOSOR40716-250G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 g
- Seq:3545
Specifications
About this item
:
:150.72 g/mol
Boiling Pt:113 °C
Density:0.866 g/cm³ (25 °C)
Flash Pt:7 °C
Storage Temperature:Ambient
UN:2924
ADR:3+8,II
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:18162-48-6
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501