Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
trans-4-Hydroxy-D-proline
trans-4-Hydroxy-D-proline 97%
trans-4-Hydroxy-D-proline 97%
APOSOR40650-5G
:
Apollo Scientific
:
trans-4-Hydroxy-D-proline 97%
APOSOR40650-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
131.131 g/mol
:
273 °C (dec.)
:
GHS07
:
Ambient
:
3398-22-9
:
C₅H₉NO₃
:
MFCD00065608