

2-(4-Aminophenyl)ethylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13472-00-9
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00008194
:136.20 g/mol
Boiling Pt:103 °C/0.3mmHg
Melting Pt:28…31 °C
Density:1.034 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Refrigerator
UN:3259
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together