Order Entry
Czech Republic
ContactUsLinkComponent
3,4-Epoxy-1-butene 97%
Flammable
Irritant
3,4-Epoxy-1-butene 97%
Catalog # APOSOR40458-5G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
3,4-Epoxy-1-butene 97%
Catalog # APOSOR40458-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :70.09 g/mol
Boiling Pt:65…66 °C
Melting Pt:–135 °C
Density:0.87 g/cm³ (25 °C)
Flash Pt:–50 °C
Storage Temperature:Refrigerator
EINECS:213-210-4
UN:1993
ADR:3,I
 :GHS02
 :GHS07
CAS Number:930-22-3
Formula:C₄H₆O
MDL Number:MFCD00005149