Methyl-3-amino-2-(benzyloxy)-5-methoxybenzoate hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1221792-96-6
:
Formula:C₁₆H₁₈ClNO₄
MDL Number:MFCD16140354
:323.78 g/mol
Melting Pt:169…171 °C
Storage Temperature:Ambient
Frequently Bought Together