Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-3-hydroxypyridazine-6-carboxylate 97%
Irritant
Methyl-3-hydroxypyridazine-6-carboxylate 97%
Catalog # APOSOR40305-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-3-hydroxypyridazine-6-carboxylate 97%
Catalog # APOSOR40305-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:189…191 °C
 :154.13 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:63001-30-9
Formula:C₆H₆N₂O₃
MDL Number:MFCD17015975