Order Entry
Czech Republic
ContactUsLinkComponent
6-Methylpyrazine-2-carboxylic acid 97%
Irritant
6-Methylpyrazine-2-carboxylic acid 97%
Catalog # APOSOR40068-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Methylpyrazine-2-carboxylic acid 97%
Catalog # APOSOR40068-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.1200 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:5521-61-9
Formula:C₆H₆N₂O₂
MDL Number:MFCD08705756