
(S)-(+)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22323-82-6
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00063239
:132.16 g/mol
Boiling Pt:82…83 °C/14mmHg
Density:1.07 g/cm³ (25 °C)
Flash Pt:80 °C
EINECS:244-910-8
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together