

2-Chlorophenylhydrazinium chloride, Technical Grade
Catalog # APOSOR3994-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:179.05 g/mol
Melting Pt:200…202 °C (dec.)
Storage Temperature:Ambient
EINECS:255-194-1
UN:3077
ADR:9+EHS,III
:GHS07
:GHS09
CAS Number:41052-75-9
Formula:C₆H₈Cl₂N₂
MDL Number:MFCD00012928