Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Dimethyl 4-boronobenzenesulphonamide 95%
Irritant
N,N-Dimethyl 4-boronobenzenesulphonamide 95%
Catalog # APOSOR3969-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N,N-Dimethyl 4-boronobenzenesulphonamide 95%
Catalog # APOSOR3969-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :229.06 g/mol
Melting Pt:140…144 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:486422-59-7
Formula:C₈H₁₂BNO₄S
MDL Number:MFCD06659849