
1-(2-Chlorophenyl)biguanide hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19579-44-3
:
Formula:C₈H₁₁Cl₂N₅
MDL Number:MFCD00053016
Melting Pt:129…134 °C
:248.12 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together