
1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10199-53-8
:
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD08271933
Melting Pt:144…145 °C
:202.21 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together