Order Entry
Czech Republic
ContactUsLinkComponent
4-(3-Methoxypropylcarbamoyl)phenylboronic acid 98%
Irritant
4-(3-Methoxypropylcarbamoyl)phenylboronic acid 98%
Catalog # APOSOR3774-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(3-Methoxypropylcarbamoyl)phenylboronic acid 98%
Catalog # APOSOR3774-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Refrigerator
CAS Number:913835-85-5
Formula:C₁₁H₁₆BNO₄
MDL Number:MFCD09027225