
4-(4-Methoxyphenyl)pyrimidin-2-amine
Catalog # APOSOR3689-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:201.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:99844-02-7
Formula:C₁₁H₁₁N₃O
MDL Number:MFCD00665912