Order Entry
Czech Republic
ContactUsLinkComponent
1-Acetyl-2-methyl-5-indolinesulphonoyl chloride
Corrosive
1-Acetyl-2-methyl-5-indolinesulphonoyl chloride
Catalog # APOSOR3671-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
1-Acetyl-2-methyl-5-indolinesulphonoyl chloride
Catalog # APOSOR3671-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.74 g/mol
Melting Pt:143…145 °C
Storage Temperature:Ambient
UN:1759
ADR:8,III
 :GHS05
CAS Number:841275-78-3
Formula:C₁₁H₁₂ClNO₃S
MDL Number:MFCD07841689