Order Entry
Czech Republic
ContactUsLinkComponent
N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide 95+%
N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide 95+%
Catalog # APOSOR360628-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acrylamide 95+%
Catalog # APOSOR360628-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.14 g/mol
Storage Temperature:Ambient
CAS Number:874363-18-5
Formula:C₁₅H₂₀BNO₃
MDL Number:MFCD22493996